Tubing, Silicone, Sani-Tech®, ULTRA-C Series
Leverancier: Saint Gobain Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ULTRA-C-012-0H ULTRA-C-020-0H ULTRA-C-030-2H ULTRA-C-040-0H ULTRA-C-058-0H ULTRA-C-062-1H ULTRA-C-062-2H ULTRA-C-062-3H ULTRA-C-062-4H ULTRA-C-078-1H ULTRA-C-078-2H ULTRA-C-078-3H ULTRA-C-078-4H ULTRA-C-078-5H ULTRA-C-093-1H ULTRA-C-093-2H ULTRA-C-093-3H ULTRA-C-125-1H ULTRA-C-125-2H ULTRA-C-125-3H ULTRA-C-125-4H ULTRA-C-125-5H ULTRA-C-156-4H ULTRA-C-156-5H ULTRA-C-187-2H ULTRA-C-187-3H ULTRA-C-187-4H ULTRA-C-250-2H ULTRA-C-250-3H ULTRA-C-250-4H ULTRA-C-250-5H ULTRA-C-312-1H ULTRA-C-312-2H ULTRA-C-312-3H ULTRA-C-312-4H ULTRA-C-312-5H ULTRA-C-375-1H ULTRA-C-375-2H ULTRA-C-375-3H ULTRA-C-375-4H ULTRA-C-375-5H ULTRA-C-500-1H ULTRA-C-500-3H ULTRA-C-500-4H ULTRA-C-500-5H ULTRA-C-500-6H ULTRA-C-625-2H ULTRA-C-625-4H ULTRA-C-625-5H ULTRA-C-750-3H ULTRA-C-750-4H
GESSULTRA-C-012-0HEA 166 EUR
GESSULTRA-C-012-0H GESSULTRA-C-020-0H GESSULTRA-C-030-2H GESSULTRA-C-040-0H GESSULTRA-C-058-0H GESSULTRA-C-062-1H GESSULTRA-C-062-2H GESSULTRA-C-062-3H GESSULTRA-C-062-4H GESSULTRA-C-078-1H GESSULTRA-C-078-2H GESSULTRA-C-078-3H GESSULTRA-C-078-4H GESSULTRA-C-078-5H GESSULTRA-C-093-1H GESSULTRA-C-093-2H GESSULTRA-C-093-3H GESSULTRA-C-125-1H GESSULTRA-C-125-2H GESSULTRA-C-125-3H GESSULTRA-C-125-4H GESSULTRA-C-125-5H GESSULTRA-C-156-4H GESSULTRA-C-156-5H GESSULTRA-C-187-2H GESSULTRA-C-187-3H GESSULTRA-C-187-4H GESSULTRA-C-250-2H GESSULTRA-C-250-3H GESSULTRA-C-250-4H GESSULTRA-C-250-5H GESSULTRA-C-312-1H GESSULTRA-C-312-2H GESSULTRA-C-312-3H GESSULTRA-C-312-4H GESSULTRA-C-312-5H GESSULTRA-C-375-1H GESSULTRA-C-375-2H GESSULTRA-C-375-3H GESSULTRA-C-375-4H GESSULTRA-C-375-5H GESSULTRA-C-500-1H GESSULTRA-C-500-3H GESSULTRA-C-500-4H GESSULTRA-C-500-5H GESSULTRA-C-500-6H GESSULTRA-C-625-2H GESSULTRA-C-625-4H GESSULTRA-C-625-5H GESSULTRA-C-750-3H GESSULTRA-C-750-4H
Tubing, Silicone, Sani-Tech®, ULTRA-C Series
Slangen Transfer Tubing
Sani-Tech® ultra-pure platinum-cured silicone tubing meets the stringent requirements of the biopharmaceutical market.
  • Maximum temperature range: –62 to +260 °C (–80 to +500 °F)
  • Imparts no taste or odour to critical stream
  • Available in 50 and 65 durometers
  • Laser etched for lot traceability
It features ultra low extractables, excellent flexibility, longer pump life and non-yellowing formulation.
Applications include sterile fill and transfers, drug delivery, cell harvest, media processing, sanitary peristaltic pump tubing, production filtration and fermentation, bioreactor process lines.
Certificaten: Meets USP Class VI and EP 3.1.9 silicone elastomer. Manufactured in an FDA registered and ISO criteria.
Bestelinformatie: Available in three different lengths.