Microscope stand, precision, Dino-Lite, RK-10A
Leverancier: Dino-Lite / IDCP B.V.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
RK-10A RK-10-FX RK-10-EX RK-10-VX RK-10-PX2
630-2517EA 327 EUR
630-2517 IDCPRK-10-FX IDCPRK-10-EX IDCPRK-10-VX IDCPRK-10-PX2
Microscope stand, precision, Dino-Lite, RK-10A
Staafstatieven
The Dino-Lite RK-10A is a sturdy and stable high-end stand that can be used with all professional Dino-Lite models. It is constructed of resilient stainless steel and lightweight aluminium and offers a very precise fine-focus adjustment as well as a quick release function.
  • Quick release button allows for fast and convenient vertical movement
  • Fine-focus adjustment knob is especially useful at high magnification
  • Adjustable safety stop to prevent the microscope from touching the specimen
  • Adjustable horizontal arm (RK-10-EX, included) has a 15 cm range with 360-degree rotation
  • Additional arm increases the effective range to observe larger objects
Extensions:
RK-10-EX: Additional horizontal arm (included with stand)
RK-10-VX: Vertical arm extension
RK-10-PX: XY positioning arm with accurate, smooth adjustment options, designed for all sorts of applications that require fine examination with a X- and Y-axis movement.
Leveringsinformatie: Supplied with HD-P1 holder and RK-10-EX horizontal arm.