Controls for POCT analyser, InnovaStar®
Leverancier: DIAGNOSTIC SYSTEMS DIASYS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
590009910062 590509910062 590009910061 590509910061
DIAS590009910062EA 0 EUR
DIAS590009910062 DIAS590509910062 DIAS590009910061 DIAS590509910061
Controls for POCT analyser, InnovaStar®
Diagnostische systemen Clinical Reagents and Kits
InnovaStar® is an automatic compact POCT analyser for the daily practice that delivers result quality comparable to measurements on laboratory analysers.
  • Barcode identification: Automatic barcode scan for reagents; optional barcode scanner for samples
  • Calibration: pre-calibrated tests
  • Data storage: 50 results
  • Measurement principle: photometric/turbidimetric
  • Method storage: Up to 19 different methods or lots
  • EDP interface: RS232 connection; USB connection via adapter