Safety goggles, 602
Leverancier: UNIVET
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
602.01.00.00 602.01.00.01
UNVT602.01.00.00EA 51.7 EUR
UNVT602.01.00.00 UNVT602.01.00.01
Safety goggles, 602
Ruimzichtbrillen
The 602 goggles have an essential frame which is ideal for each type of fit.
  • Adjustable elastic fabric headband
  • Polycarbonate lens
  • Indirect ventilation system
  • OTG (worn over prescription glasses)
These goggles are used for mechanical works in good visibility conditions.
Wearing these goggles for prolonged period of time will not create distortion or fatigue.
Weight: 73 g