Swabs for surface validation, CONSTIX® SV-2 / SV-3
Leverancier: CONTEC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SV-2 SV-3
CONTSV-2EA 18.7 EUR
CONTSV-2 CONTSV-3
Swabs for surface validation, CONSTIX® SV-2 / SV-3
Wattenstaafjes Wattenstaafjes voor reiniging
Sealed knitted polyester swabs for surface cleaning validation.
  • Heads are constructed of a double-layer knitted polyester fabric that has been processed to minimise background TOC contribution
  • Heads are thermally bonded to a notched PP handle to minimise handling and improve sample insertion into vials
  • Polyester heads are constructed without adhesives or binders
  • Compatible with most common solvents, such as alcohols and ketones
  • Swabs can be sterilised by most common methods
  • Small put-up ensures accurate testing with little waste
SV-2: The round head and flexible handle of SV-2 allows access to recessed surfaces for easier contamination retrieval or sampling.
SV-3: The large paddle head provides a broader surface coverage for sampling and contamination retrieval in critical applications.
Verpakking: Each pack contains five swabs packed inside a cleanroom grade re-closable bag.
Opgelet: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904