Cleaning cloths, Unitex®
Leverancier: Zellstoff-Vertriebs-GmbH & Co. KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11148-01 10148-01
ZVGA11148-01EA 157 EUR
ZVGA11148-01 ZVGA10148-01
Cleaning cloths, Unitex®
Schoonmaakdoekjes Herbruikbare reinigingsdoeken Doeken voor algemeen gebruik
These Unitex® cleaning cloths are excellent for cleaning and degreasing surfaces, in conjunction with cleaning agents like silicone removers, water based cleaners or water. Features special manufacturing process in which polyester and pulp fibres are woven together by hydro power.
  • Tear resistant
  • High strength