Peaked caps
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
113-9236EA 0 EUR
113-9236 113-9237 113-9238 113-9239
Peaked caps
Wegwerpkleding
These disposable caps, with peak, are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
  • Free from natural rubber latex
  • Available in white and blue
Weight: 40 g/m² (top), 100 g/m² (peak)
Verpakking: 100 pieces per polybag and 1000 pieces per carton.