Vertical laminar flow cabinets, DLF series
Leverancier: CleanAir by Baker
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
NP0024360 NP0037460 NP0024460 NP0024560 NP0024660 NP0037360
135-0078EA 0 EUR
135-0078 CLEANP0037460 CLEANP0024460 CLEANP0024560 CLEANP0024660 CLEANP0037360
Vertical laminar flow cabinets, DLF series
Zuurkasten en behuizingen Kasten met laminaire stroming
CleanAir by Baker DLF series is a high quality vertical laminar flow workbench range for handling non-hazardous materials that require a clean working environment. DLF PCR Series is designed to perform PCR. These cabinets ambient air entering the work area, ensures product protection.
  • Microprocessor flow control
  • Smooth interior for efficient and easy cleaning
  • Control control system with alarms and parameters for work mode, UV light, gas valve
  • Epoxy coated steel interior and exterior (1,5 to 2 mm)
  • Stainless Steel worktop
  • Sliding window (safety glass)
  • Controllable by programmable clock (on/off) or timer (off)
  • HEPA filtered vertical laminar airflow (downflow)
DLF series have been specially designed to work in pharmaceutical industry, biotechnology, pharmacies, hospitals, university laboratories, tissue culture and electronic laboratories.
Certificaten: ISO Class 5, GMP Class A, 2006/42/EC, 2004/108/EC, 2006/95/EC, EN 61010-01.