Visitor coats, antistatic, PP
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
113-9271EA 0 EUR
113-9271 113-9270
Visitor coats, antistatic, PP
Wegwerpkleding
Disposable visitor coats made from PP.
  • Front closure with five buttons
  • Korean collar and antistatic
  • Latex-free elastic wrist
Weight: 35 g/m²
Verpakking: One per bag and 50 per carton.
Order Now
Specificaties
Environmentally Preferable
Sluitingstype Met knoppen
Geslacht Uniseks
Kledingstuk toepassing Laboratory,Industrial