Heptafluorobutyric acid 0.5 M for ion pair chromatography, Supelco®
Leverancier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
49540-100ML-FEA
324
EUR
49540-100ML-F
Heptafluorobutyric acid 0.5 M for ion pair chromatography, Supelco®
Heptafluorobutyric acid
Formule:
C₄HF₇O₂ MW: 214,04 g/mol |
MDL Nummer:
MFCD00004171 CAS nummer: 375-22-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...