1-(Trimethylsilyl)imidazole silylation reagent, for GC derivatisation
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: TSIM, N-(Trimethylsilyl)imidazole, N-(Trimethylsilyl)-1H-imidazole, TMSI, 1-(Trimethylsilyl)-1H-imidazole
701310.110 701310.201 701310.510
MANA701310.110EA 64 EUR
MANA701310.110 MANA701310.201 MANA701310.510
1-(Trimethylsilyl)imidazole silylation reagent, for GC derivatisation
1-(Trimethylsilyl)imidazole
TSIM is the strongest hydroxyl silylator; reagent of choice for carbohydrates and most steroids (even highly hindered steroids).
  • Silylation can be applied on many compounds
  • Silylation reagents are easily prepared
  • TSIM selectively derivatizes compounds with different functional groups
The reagent is unique in that it reacts quickly and smooth with hydroxyl (even tert. OH) and carboxyl groups, but not with amines. This characteristic makes TSIM particularly useful in multi-derivatization schemes for compounds with different functional groups, which are to be derivatized differently (e.g., -O-TMS / -N-HFB derivatives of catecholamines). Recommended applications are the derivatizatzion of alcohols, phenols, organic acids, steroids, hormones, glycols, nucleotides and narcotics.
Formule: C₆H₁₂N₂Si
MW: 140.26 g/mol
Kookpunt: 94–96 °C (760 mm Hg)
Smeltpunt: -42
Dichtheid: 0.961 g/cm³ (d20°/4°)
Vlampunt: 61
MDL Nummer: MFCD00005280
CAS nummer: 18156-74-6