Magnifier lamps, varioLED+
Leverancier: ESCHENBACH OPTIK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
27773 27776
630-3079EA 889 EUR
630-3079 ESCH27776
Magnifier lamps, varioLED+
Vergrootglazen en loepen
The varioLED+ magnifying lamp provides uniformly bright illumination of the work surface without casting shadows via ring lighting thanks to 84 long-lasting, high-performance LEDs.
  • Illumination levels (ring lighting, 200 mm): 6000 lx, 3500 lx, 2400 lx
  • Luminous flux (max.): 1000 lm
  • Colour temperature: approx. 6500 K
  • Lens diameter: 132 mm
  • Energy efficiency class: A
Lamp is equipped with an aspherical PXM® lightweight lens with a cera-tec® hard coating for sharp, distortion-free images, available in 1,9× (3,55 dpt) or 2,5× (6,0 dpt) magnification. The magnifier head features 6 joints to ensure optimum positioning. A clearly-structured operation panel includes separate buttons for on/off, switching between ring and segment lighting, and dimming (40%, 60%, 100%). An easily removable hinged lens cover also serves as a dust protector. The light contains built-in LED lamps, which cannot be replaced.