Safety welding goggles, Ultrasonic Flip-up
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9302.045 9302.043
UVEX9302.045EA 53.1 EUR
UVEX9302.045 UVEX9302.043
Safety welding goggles, Ultrasonic Flip-up
Ruimzichtbrillen
Wide-vision welding safety goggles feature a sporty design, and are also suitable as overspecs.
  • Perfect colour recognition thanks to Uvex sunglare filters
  • New filter technology combines UV 400 and IR protection
  • Filter extremely scratch-resistant on both sides, minimizing burns due to welding sparks (uvex infradur)
  • No problems with pressure when worn for long periods, due to a combination of hard and soft components
The combination of hard and soft material components used in the uvex ultrasonic pairs high mechanical strength with optimum wearability. This wide-vision goggle can be worn for long periods without causing any pressure problems. The flexible soft component allows it to adapt to each individual face for a snug fit, ensuring comprehensive side protection around the eye area, even when worn over prescription spectacles.
The uvex ultrasonic offers outstanding ventilation, ensuring a pleasant interior environment that does not cause irritation. They also features a sporty design, panoramic field of vision, and quick and easy lens replacement. The welding safety version of the goggle has a practical flip-up lens that is very easy to replace.
Certificaten: EN 166; EN 169; EN 170