Chinese ink, zwart, Q Path® stain for histology
Leverancier: VWR Q-Path Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
01816200.EA 83.4 EUR
01816200.
Chinese ink, zwart, Q Path® stain for histology
Chinese ink
Q Path Inks for Pathology are inks for marking and identifying histological samples without altering the tissues.
  • Very long lasting non removable ink
  • Product ready for use
Usage
1. Shake the bottle well before opening
2. The ink is applied using a paintbrush or a swab
3. Close the bottle properly after use
Composition: Water, pigment and glycerine.
Opgelet: These reagents must only be handled by professionals