Flat mops
Leverancier: MICRONOVA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MZFM7-14 MZFM7-14IR MZFM7-18 MZFM7-18IR MZFM7-9
MICRMZFM7-14EA 36.4 EUR
MICRMZFM7-14 MICRMZFM7-14IR MICRMZFM7-18 MICRMZFM7-18IR MICRMZFM7-9
Flat mops
Moppen
These brick-style, flat sponge mop heads are excellent for applying disinfectant in pharmaceutical production or aseptic processing applications.
  • Easily clean large surfaces
  • Heavy-duty construction offers additional resistance to textured floors and aggressive cleaning methods
  • Fully padded mop heads can be laundered and reused several times
Ideal for cleaning large surface areas as well as reaching shelving, doorjambs, and corners.
Mop heads feature heavy-duty construction with PolyTex™ textured polyester material, PolyTex™ material combined with MegaTex™ textured non-woven polyamide fabric strip, or PolyTex™ material combined with microfibre fabric strip. PolyTex™/MegaTex™ mop heads offer additional resistance to textured floors and more aggressive action for spills and heavy soils and these heads allow faster pick up of moisture in standard clean/disinfect applications such as hospitals and animal labs.
Bestelinformatie: All components must be purchased separately. An electropolished stainless steel quick-disconnect adapter is required to attach mop head to an electropolished stainless steel extension handle.