Organic reference standard, 3-(3-Hydroxyphenyl)propionic acid, analytical standard, Supelco®
Leverancier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
91779-10MG
91779-50MG
91779-10MGEA
39.6
EUR
91779-10MG
91779-50MG
Organic reference standard, 3-(3-Hydroxyphenyl)propionic acid, analytical standard, Supelco®
Standaarden
Organische standaarden
3-(3-Hydroxyphenyl) propanoic acid is one of the major metabolites of ingested caffeic acid and of the phenolic degradation products of proanthocyanidins (the most abundant polyphenol present in chocolate) by the microflora in the colon. 3-(3-Hydroxyphenyl) propanoic acid is suspected to have antioxidants properties and is actively absorbed by the monocarboxylic acid transporter (MCT) in intestinal Caco-2 cell monolayers.