Organic reference standard, Allyl sulfide, analytical standard, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
40647-1ML 40647-5ML
40647-1MLEA 48.6 EUR
40647-1ML 40647-5ML
Organic reference standard, Allyl sulfide, analytical standard, Supelco®
Standaarden Organische standaarden
An organosulphur compound from garlic that inhibits chemically-induced carcinogenesis in experimental animals. Competitive inhibitor of Cytochrome P450 2E1 (CYP2E1) that, in turn, blocks the activation of several chemcal carcinogens.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5adb1b721ba8c50e78bd349adc075dbd71351673&vid=5adb1b721ba8c50e78bd349adc075dbd71351673&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf19be99cf0acb4a2a1a767362550f9e3e1ffab3&vid=bf19be99cf0acb4a2a1a767362550f9e3e1ffab3&prid=web&clid=SAPDAM