Phytochemical standard, Dihydromyricetin, analytical standard, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
42866-10MGEA 148 EUR
42866-10MG
Phytochemical standard, Dihydromyricetin, analytical standard, Supelco®
Standaarden Organische standaarden
Dihydromyricetin (ampelopsin) is a flavanonol with antioxidant and anti-cancer activity, found to have anti-alcohol intoxication effects. Its anti-alcohol effects appear to be by its actions as a positive modulator of GABA-A receptors at the benzodiazepine site.