Organic reference standard, Dulcoside A, analytical standard, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
90378-10MGEA 883 EUR
90378-10MG
Organic reference standard, Dulcoside A, analytical standard, Supelco®
Standaarden Organische standaarden
Dulcoside A is a steviol glycoside extracted from the leaves of Stevia rebaudiana bertoni plant.
It has applications as a natural sweetener.