Glucose assay reagent, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
G3293-50ML G3293-20ML
G3293-50MLEA 157 EUR
G3293-50ML G3293-20ML
Glucose assay reagent, Supelco®
Testsystemen voor voeding en drank
Activity <0,05% resulting from cross-reactivity with the following substrates: fructose, dextran, sedoheptulose, arabinose, galactose, xylose, trehalose, mannose, sorbitol, fucose or ribose.
For the quantitative determination of glucose in food and other materials utilizing the hexokinase-glucose 6-phosphate dehydrogenase enzymatic assay. Reconstitute with deionised water using volume indicated by package size (20 or 50 ml).
Verpakking: 20, 50 ml in serum bottle.
Order Now
Specificaties
Omschrijving Glucose Assay Reagent
Environmentally Preferable