Supelco® Glucose Oxidase/Peroxidase Reagent
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
G3660-1CAPEA 46.3 EUR
G3660-1CAP
Supelco® Glucose Oxidase/Peroxidase Reagent
Testsystemen voor voeding en drank
Glucose oxidase/peroxidase reagent along with O-Dianisidine was used for the determination of blood and urinary glucose, especially the presence of fructose for both research and clinical purpose.
It may be used for determining glucose in plasma or serum. It may also be used in the estimation of blood glucose as a function of the oxidative coupling system or of the oxygen affinity of the oxygen acceptor chromogen.
Verpakking: One cap in glass bottle.