Starch (GO/P) assay kit, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
STA20-1KTEA 297 EUR
STA20-1KT
Starch (GO/P) assay kit, Supelco®
Testsystemen voor voeding en drank
This kit is for the quantitative, enzymatic determination of starch in food and other materials. Starch (GO/P) assay kit was used to determine the starch content, during an experimental study done in order to investigate the composition and structure of sorghum spent grains (SSG), which is a brewery co-product and is reported to have potential applications thereby adding value to the product.
It was also used in starch extraction and quantification, during investigating the importance of OsGA2ox5 in GAs homeostasis, development, gravity responses and stress tolerance in rice.Furthermore, it also found application in being used in analysing factors affecting polyhydroxybutyrate accumulation , which are basically linear polyesters produced by bacteria as a carbon store, in mesophyll cells of sugarcane and switchgrass.
Starch (GO/P) assay kit was used to determine the starch content, during an experimental study done in order to investigate the composition and structure of sorghum spent grains (SSG), which is a brewery co-product and is reported to have potential applications thereby adding value to the product. It was also used in starch extraction and quantification, during investigating the importance of OsGA2ox5 in GAs homeostasis, development, gravity responses and stress tolerance in rice.
It also found application in being used in analysing factors affecting polyhydroxybutyrate accumulation, which are basically linear biodegradable polyesters produced by bacteria as a carbon store, in mesophyll cells of sugarcane and switchgrass.
The hydrolysis of starch to glucose is catalysed by α-amylase and amyloglucosidase. Glucose is oxidised to gluconic acid and hydrogen peroxide by glucose oxidase. Hydrogen peroxide reacts with o-dianisidine in the presence of peroxidase to form a colored product. Oxidised o-dianisidine reacts with sulphuric acid to form a more stable coloured product.The intensity of the pink color measured at 540 nm is proportional to the original glucose concentration.
This kit is for the quantitative, enzymatic determination of starch in food and other materials. The hydrolysis of starch to glucose is catalysed by α-amylase and amyloglucosidase. Glucose is oxidised to gluconic acid and hydrogen peroxide by glucose oxidase. Hydrogen peroxide reacts with o-dianisidine in the presence of peroxidase to form a colored product. Oxidised o-dianisidine reacts with sulphuric acid to form a more stable colored product. The intensity of the pink color measured at 540 nm is proportional to the original glucose concentration.