Supelco® Organic Reference Standard, Ursolic acid
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
89797-25MG-F 89797-5MG-F
89797-25MG-FEA 472 EUR
89797-25MG-F 89797-5MG-F
Supelco® Organic Reference Standard, Ursolic acid
Standaarden Organische standaarden
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2c331e118415661c80241b44a005dab00c3fdfa&vid=d2c331e118415661c80241b44a005dab00c3fdfa&prid=web&clid=SAPDAM
Order Now
Specificaties
Synonym(s) 3β-Hydroxy-12-ursen-28-ic acid
Environmentally Preferable
M-Clarity quality level MQ100
CAS No. 77-52-1
MDL No. MFCD00009621
Moleculair gewicht 456,70
Smeltpuntapparaat 292 °C (dec.) (lit.)
Shipping conditions STD
Storage conditions 2...30 °C