Kettle brushes
Leverancier: MICRONOVA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BRDB-41 BRWH-41 BRWH-42
MICRBRDB-41EA 126 EUR
MICRBRDB-41 MICRBRWH-41 MICRBRWH-42
Kettle brushes
Borstels
The kettle brush is an oblong-shaped brush with a flare head design and polyester bristles, that is well suited for an all-around, thorough cleaning of small tanks, carboys and other production equipment.
  • Lightweight, durable brush with optional fabric covers
  • Made from FDA approved, BPA-free base resins
  • FDA approved for indirect food contact
  • Features three ACME threaded handle attachments, one at 90° and two angled on either side
  • Optional covers and handles sold separately
  • Autoclavable, with base down
Attaches to Micronova's STA series, fixed and extendible length handles to reach the bottom of large process tanks and kettles.