Cleanroom barrier tapes
Leverancier: MICRONOVA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BT100-3RE BT200-3YE
MICRBT100-3REEA 81.1 EUR
MICRBT100-3RE MICRBT200-3YE
Cleanroom barrier tapes
Tape
Tapes made of high visibility, non-adhesive polyethylene film. Ideal for blocking restricted areas and recommending caution in controlled environments.
  • Text is in English and Spanish (2" hgh lettering)
  • Yellow or red film with bold black print
  • Wound on a 76 mm (3") plastic core
These tapes help prevent accidents by increasing visibility of hazards and can be tied, stapled or nailed in place.
Length: 304,8 m