Safety goggles, 625
Leverancier: UNIVET
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
625.03.00.00 625.03.01.05
625-0000EA 0 EUR
625-0000 UNVT625.03.01.05
Safety goggles, 625
Ruimzichtbrillen
The 625 goggle is equipped with a wide panoramic lens for a better side protection, an adjustable elastic band and a soft frame.
  • Extremely lightweight
  • Lens with anti-fog Vanguard PLUS coating
  • Panoramic, wraparound lens for a better side protection
  • Adjustable headband and soft frame effectively reduce pressure on the head
  • Frame also protects against drops, splashes and larger dust particles
Available with clear PC lens for UV and impact protection or with G15 solar lens for UV protection and perfect colour recognition (TSR).
Certificaten: EN 166, EN 170