Cell culture flasks, Nunc™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
153732 156340 156367 156472 156499 156502 156800 159910 159920 178883 178905 169900 159926 178983 159933 132707 132708 132703 132704 132705 132706 178885 178985 156505 159934 132097 132098
734-2046EA 115 EUR
734-2046 734-2063 734-2064 734-2065 734-2066 734-2067 734-2069 734-2167 734-2168 734-2129 734-2131 734-2104 734-1190 734-1340 734-1337 734-2285 734-2286 734-2281 734-2282 734-2283 734-2284 392-0390 392-0391 392-0389 734-1338 734-2608 734-2609
Cell culture flasks, Nunc™
Kolven Celcultuurflessen
These Nunc™ cell culture flasks are designed for culture consistency, cell health, and reproducibility. Choose from a variety of surfaces and sizes with culture areas ranging from 25 to 500 cm² to suit your specific applications and cell types.
  • Nunclon Delta certification including testing on four different cell types is provided in each case
  • Barcoded 175 cm² flasks (*) with large Code 128 are compatible with automation
  • Sterilized by gamma irradiation and certified to be non-pyrogenic
Flasks treated with the proprietary Nunclon™ Delta surface treatment ensure consistent cell growth and maximum adhesion for a broad range of cell types. Nunc™ non-treated flasks are made with high-quality, optically clear virgin polystyrene with a hydrophobic surface ideal for suspension cell culture and useful for a variety of biochemical assays.
The ergonomic design of Nunc™ EasYFlask with 1/3-turn cap enables one-handed operation and avoids wrist strain. The angled, extra-wide neck provides easier access to growth surface with cell scrapers or pipettes. Molded and printed graduations help enable easy and quick measurement of growth media. Nunc™ standard flasks are designed with a straight neck and barcoding option for automation cell culture.
Options of solid or filtered cap for adequate gas exchange. Solid caps are vent/close caps with 'Y' marks that allow easy visual verification of venting position while promoting sufficient gas exchange. Filter caps ensure constant gas exchange and constant airflow while minimizes chances of contamination.