Rotoren voor 5420, eppendorf
Leverancier: EPPENDORF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5495400.001 5495404.007
521-2722EA 690 EUR
521-2722 521-2723
Rotoren voor 5420, eppendorf
Centrifuge Rotoren
Eppendorf Rotors for Centrifuge 5420, precision for diverse applications. Choose from various capacities and speeds to tailor your centrifuge setup. Accessorize with adaptors and buckets, ensuring optimal sample processing in molecular biology and beyond.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 min)
  • Accessories (adaptors and buckets) further customize your centrifuge setup
  • Aerosol-tight options available
Elevate your centrifugation capabilities withMore Product Information