Vanadium standard for ICP, TraceCERT®, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
18399-100ML 44712-100ML
18399-100MLEA 42.2 EUR
18399-100ML 44712-100ML
Vanadium standard for ICP, TraceCERT®, Supelco®
Standaarden Kationische standaarden Vanadium standaarden
Certificaten: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034