Redox standards, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
78855-500ML 80138-500ML 87783-500ML 13198-500ML 04887-500ML 40661-500ML
78855-500MLEA 127 EUR
78855-500ML 80138-500ML 87783-500ML 13198-500ML 04887-500ML 40661-500ML
Redox standards, Supelco®
Standaarden Redox- en ORP-standaarden
The values are the potential of a platinum electrode versus an Ag/AgCl reference in 3M potassium chloride.