Micro particles based on melamine resin, carboxylate-modified FITC-marked, analytical standard, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
88486-5ML-F 90287-5ML-F
88486-5ML-FEA 0 EUR
88486-5ML-F 90287-5ML-F
Micro particles based on melamine resin, carboxylate-modified FITC-marked, analytical standard, Supelco®
Standaarden Standaarden voor particelgrootte
For every lot exact values of particle size and standard deviation are determined with an accuracy of 0,01 μm using a Coulter Multisizer.
Surface coverage: Reactive groups (COOH) >0,1 mmol/g polymer.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e3c47513ba429429adc564aae5912e3aa0707a0c&vid=e3c47513ba429429adc564aae5912e3aa0707a0c&prid=web&clid=SAPDAM