Micro particles based on melamine resin FITC-marked, analytical standard, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
90305-10ML-F 90305-5ML-F 94050-10ML-F
90305-10ML-FEA 603 EUR
90305-10ML-F 90305-5ML-F 94050-10ML-F
Micro particles based on melamine resin FITC-marked, analytical standard, Supelco®
Standaarden Standaarden voor particelgrootte
For every lot, exact values of particle size and standard deviation are determined with an accuracy of 0,01 μm using a Coulter Multisizer.
Fluorescence: λex 490 nm; λem 525 nm
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80d3a9e078adf79a26ec3b8e2da41c77265a444f&vid=80d3a9e078adf79a26ec3b8e2da41c77265a444f&prid=web&clid=SAPDAM