Funnel, stainless steel
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9604-0008 9604-0003 9604-0005 9604-0001
221-0283EA 48.8 EUR
221-0283 BURK9604-0003 BURK9604-0005 BURK9604-0001
Funnel, stainless steel
Trechters Filtertrechters Filtertrechters
This funnel can be used to pour and transfer free-flowing powders, liquids and slightly viscous media easily.
  • Made of stainless steel AISI 304 (1.4301)
  • An air channel on the discharge tube ensures a complete, unobstructed flow
  • Handle ensures easy handling
Depending on the diameter, this pouring aid is suitable for dispensing applications, from small amounts in laboratories to large quantities in industry.
The sturdy stainless-steel funnel can be used in a wide variety of applications.