Palette knife spatula
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5386-0012 5386-0010
BURK5386-0012EA 10.8 EUR
BURK5386-0012 BURK5386-0010
Palette knife spatula
Spatels
Similar to a pallet or dispensing chemist's spatula, this high quality palette knife spatula is suitable for applying or distributing viscous media such as pastes and creams, or for spreading them smoothly.
  • Made entirely from stainless steel (1.4301)
  • No edges or crevices: Facilitates cleaning and prevents product residue and cleaning agents from accumulating
  • Long, narrow, high-gloss polished blade facilitates precise work
  • Sturdy handle fits comfortably in the hand
The laboratory spatula is suitable for use in chemical and pharmaceutical applications.