3-(N-Ethyl-3-methylanilino)-2-hydroxypropanesulfonic acid sodium salt, for enzymic spectrophotometric det. of H2O2, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E8631-1G E8631-5G
E8631-1GEA 225 EUR
E8631-1G E8631-5G
3-(N-Ethyl-3-methylanilino)-2-hydroxypropanesulfonic acid sodium salt, for enzymic spectrophotometric det. of H2O2, Supelco®
Standaarden Organische standaarden
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af864b78ea720f2c537f2e36d645e9a85132ea31&vid=64936116f27df5b877c83e88ab93955f398ab370&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13ed7d91dfbdaf8dc0c55c550fc5f4c636b4dbde&vid=119c8a3d7e6bd7e134deabb587cfddddca054e2f&prid=web&clid=SAPDAM