Sigma Pseudo (TM) Corpse Scent, Formulation I, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P4304-20ML P4304-4X1ML P4304-8X1ML
P4304-20MLEA 270 EUR
P4304-20ML P4304-4X1ML P4304-8X1ML
Sigma Pseudo (TM) Corpse Scent, Formulation I, Supelco®
Standaarden Organische standaarden
Canine training aids for the detection of corpses.
Sigma pseudo™ corpse scent has been used as a test material for determining the components present in it using techniques such as a direct injection, headspace solid phase microextraction (SPME), purge and trap, and thermal desorption followed by gas chromatography coupled with mass spectrometry.
Opgelet: Storage temperature: 2 to 8 °C