IQ/OQ/PQ Kit for Ion Chromatography, certified reference material, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
12674-1KTEA 334 EUR
12674-1KT
IQ/OQ/PQ Kit for Ion Chromatography, certified reference material, Supelco®
Standaarden Anionische standaarden Anionische standaarden met meerdere elementen
IQ/OQ/PQ-Kit for the qualification of ion chromatography systems. Suitable for the verification of linearity, repeatability and carry-over.
Certificaten: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034
Verpakking: Contains six certified bromide standards (5, 10, 20, 50, 100 and 1000 mg/kg; 100 ml each). The exact contents, traceability and uncertainty are given in a detailed certificate.