Avantor® Partisil PAC, HPLC Columns
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P10PAC-250AEA 1150 EUR
P10PAC-250A P5PAC-250A P5PAC-10C5 P10PAC-250AF
Avantor® Partisil PAC, HPLC Columns
Kolommen voor chromatografie
Avantor® Partisil® was one of the first commercially available irregular silicas with a large surface area giving it a high loading capacity.
  • Easy to use
  • Irregular porous silica
Avantor® Partisil® PAC is a polar amino cyano bonded phase with secondary amine groups for good thermal and chemical stability. Fast equilibration enables multiple separation mechanisms, including adsorption, reversed-phase and weak anion-exchange, to be used. The phase is particularly suitable for carbohydrate separations.
Opgelet: * Use with freestanding holder HI-161 andMore Product Information