Avantor® Partisil Silica, HPLC Columns
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P5-250AEA 1220 EUR
P5-250A P10-250A P5-150A P5-200A P5-250AS P10-150AF P10-250AF P5-100A P5-250AF
Avantor® Partisil Silica, HPLC Columns
Kolommen voor chromatografie
Avantor® Partisil® was one of the first commercially available irregular silicas. A large surface area gives it a high loading capacity. Avantor® Partisil® ion-exchange materials are widely referenced and remain one of the most popular choices for analysts.
  • Irregular porous silica
  • 5 and 10 µm particle sizes
  • Hichrom high efficiency