Laboratory coats, PP
Leverancier: Hubei Orient International
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
114-9025EA 102 EUR
114-9025 114-9024 114-9023 114-9012 114-9014 114-9015 114-9020 114-9022 114-9013 114-9016 114-9017 114-9018 114-9019 114-9021 114-9026 114-9027 114-9028
Laboratory coats, PP
Wegwerpkleding
These disposable laboratory coats with shirt neck made from PP and are ideal for non critical applications.
  • Elasticated or Velcro wrist
  • Press stud fastening (four/five snaps)
Weight: 25 to 55 g/m²