Overalls without hood or feet cover, PP
Leverancier: Hubei Orient International
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HOIT110310EA 166 EUR
HOIT110310 HOIT110694 HOIT110437 HOIT109739 HOIT114044 HOIT114045 HOIT110398 HOIT109740 HOIT115308
Overalls without hood or feet cover, PP
Wegwerpkleding
These disposable overalls are made from non woven PP. Ideal for non critical applications in industrial, clinical or food processing environments.
  • Without hood and feet cover
  • Excellent protection against a wide range of hazards and nuisances
Weight: 22 to 55 g/m²
Colour: White