VWR® ARISTAR®, Multi-Element Standards, Anionic for Ion Chromatography
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
87037.260EA 258 EUR
87037.260 87038.260 87039.260 87040.180
VWR® ARISTAR®, Multi-Element Standards, Anionic for Ion Chromatography
Standaarden Anionische standaarden Anionische standaarden met meerdere elementen
  • Traceable to NIST
  • Manufactured and tested in ISO Guide 34 / ISO 17025 accredited facilities
Leveringsinformatie: Supplied with a Certificate of Analysis.