VWR® ARISTAR®, Multi-Element Standards, Cationic for Ion Chromatography
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
87041.180EA
130.2
EUR
87041.180
87042.180
VWR® ARISTAR®, Multi-Element Standards, Cationic for Ion Chromatography
Standaarden
Kationische standaarden
Standaarden met meerdere elementen
- Traceable to NIST
- Manufactured and tested in ISO Guide 34 / ISO17025 accredited facilities
- Supplied with a Certificate of Analysis
Ion chromatography cation multi-element standard solution VII, ARISTAR® (100 mg/L: Li, Na, K, NH₄, Mn, Ca, Mg, Sr, Ba in 0,1% HNO₃)