Exsil Silica, HPLC Columns
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EX-5-250AEA 420 EUR
EX-5-250A EX-10-250P EX80-5-250A
Exsil Silica, HPLC Columns
Kolommen voor chromatografie
Exsil HPLC columns are a useful alternative to the classical spherical HPLC silicas.
  • Spherical porous silica
  • 80 and 100 Å pore sizes
  • Competitively priced
  • Hichrom high efficiency
Exsil 100, the standard material, is supplied in a range of particle sizes and bonded phases to cover a wide range of applications. Exsil 80 is a higher surface area, complementary range of materials.