Avantor® Hichrom, ValvTool
Leverancier: VWR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Avantor® Hichrom
HI-199EA 71.1 EUR
HI-199
Avantor® Hichrom, ValvTool
Moersleutels
This Valvtool is a uniquely designed slotted wrench which provides easy access to many hard-to-reach areas.
It enables fittings, sample loops and most other ¼" stainless steel and PEEK HPLC fittings to be more easily tightened or loosened.