Avantor® Hichrom, Frits, Cap Style
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HI-305EA 73 EUR
HI-305 HI-301 HICHHI-302 HICHHI-303
Avantor® Hichrom, Frits, Cap Style
Filters Voorkolom filters en frits
These cap style frits used to preserve the packaging material within the column itself.