Diluid ABACUS Haematology, J.T.Baker®
Leverancier: Avantor
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3430.9010EA
24.34
EUR
3430.9010
3430.9020
Diluid ABACUS Haematology, J.T.Baker®
Diluid
Opgelet: In vitro diagnostic medical devices only for professional use.
![]() |
Specification Test Results
Background PLT (x10E9/L) | max. 5 |
Background RBC (x10E12/L) | max. 0.02 |
Background WBC (x10E9/L) | max. 0.3 |
Conductivity (mS) | 16.00 - 21.00 |
Osmolality (mOsm/kg) | 325 - 335 |
Performance Test | passes test |
pH at 25°C | 6.90 - 7.10 |
Total Bacterial Growth (CFU/100ml) | max. 5 |