Amberlite® XAD-7 HP, Pore size: 300-400 Å (mean) 20 - 60 mesh synthetic adsorbent resin, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Amberlite® XAD7HP
M-Clarity™ quality level = MQ100
XAD7-1KG XAD7-100G XAD7-500G
XAD7-1KGEA 258 EUR
XAD7-1KG XAD7-100G XAD7-500G
Amberlite® XAD-7 HP, Pore size: 300-400 Å (mean) 20 - 60 mesh synthetic adsorbent resin, Supelco®
Amberlite® XAD-7 HP
Nonionic macroreticular resin that adsorbs and releases ionic species through hydrophobic and polar interactions; usually used under isocratic conditions.
Weakly polar adsorbent resin for compounds up to 60,000 MW: insulin, fulvic and humic compounds, dry waste, organic removal and recovery, and antibiotic recovery.
MDL Nummer: MFCD00132705
CAS nummer: 37380-43-1