Cyanine 647 succinimidyl ester cyanine dye
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
90025-10MG 90025-1MG
90025-10MGEA 1840 EUR
90025-10MG 90025-1MG
Cyanine 647 succinimidyl ester cyanine dye
Cyanine succinimidyl ester
Biotium offers high quality amine-reactive succinimidyl ester (SE) dyes used to label proteins, oligonucleotides, and nucleic acids.
  • Cyanine 647 is structurally identical to Cy®5 from GE Healthcare.
  • λEx/λEm (water) = 650/665 nm
  • Dark blue solid soluble in DMF and DMSO
  • Store at -20°C and protect from moisture and light
We offer Cyanine 555 (Cy®3), Cyanine 647 (Cy®5), Cyanine 680 (Cy®5.5), and Cyanine 750 (Cy®7) as high quality succinimidyl esters.
Also see our CF® Dye Succinimidyl Esters, available with more than 20 bright and photostable CF® Dyes.
Cy3, Cy5, Cy5.5, and Cy7 are registered trademarks of GE Healthcare.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b6328352525fbf4e78a11328e0f93c7ad76d8fdb&vid=2d6d651b00ecc1f0dae9341eb8a7bf33d1f3892a&prid=web&clid=SAPDAM
Order Now
Specificaties
Environmentally Preferable