Amberlite® IR-120 (Na) ion-exchange resin, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
224359-1KG 224359-250G
224359-1KGEA 193 EUR
224359-1KG 224359-250G
Amberlite® IR-120 (Na) ion-exchange resin, Supelco®
Amberlite® IR-120 (Na)
Amberlite® IR120 Na+ form may be used in separation medium for TLC, paper chromatography and LPLC.
MDL Nummer: MFCD00132707
CAS nummer: 9002-23-7